MilliporeSigma
  • Optical turn-on sensor based on graphene oxide for selective detection of D-glucosamine.
Optical turn-on sensor based on graphene oxide for selective detection of D-glucosamine.
Analytical chemistry (2012-06-05)
Rumei Cheng, Yong Liu, Shengju Ou, Yaqiong Pan, Shu Zhang, Hao Chen, Liming Dai, Jia Qu
ABSTRACT
By incorporating the well-known fluorophore 8-aminoquinoline into graphene oxide, we have successfully prepared a turn-on fluorescent sensor capable of specific detection of D-glucosamine with a high selectivity and sensitivity. This methodology provides a new concept for the design and development of highly selective and sensitive turn-on optical sensors for selective detection of aminosaccharides and many other biomolecules.
Sigma-Aldrich
8-Aminoquinoline, 98%