MilliporeSigma
  • Complete genome sequence of the fenitrothion-degrading Burkholderia sp. strain YI23.
Complete genome sequence of the fenitrothion-degrading Burkholderia sp. strain YI23.
Journal of bacteriology (2012-01-26)
Jong Sung Lim, Beom Soon Choi, Ah Young Choi, Kyung Duk Kim, Dong In Kim, Ik Young Choi, Jong-Ok Ka
ABSTRACT
Burkholderia species are ubiquitous in soil environments. Many Burkholderia species isolated from various environments have the potential to biodegrade man-made chemicals. Burkholderia sp. strain YI23 was isolated from a golf course soil and identified as a fenitrothion-degrading bacterium. In this study, we report the complete genome sequence of Burkholderia sp. strain YI23.
Product Description
Supelco
Fenitrothion, PESTANAL®, analytical standard