MilliporeSigma
  • Efficient double glycoconjugation to naturalize high molecular weight disperse dyes.
Efficient double glycoconjugation to naturalize high molecular weight disperse dyes.
Carbohydrate research (2011-12-06)
Roberto Bianchini, Massimo Rolla, Jalal Isaad, Giorgio Catelani, Lorenzo Guazzelli, Massimo Corsi, Marco Bonanni
ABSTRACT
Commercially available Disperse Orange 29 (1a) and Disperse Red 1 (2a) were elaborated to glycoconjugated species, following a new version of a previously-described 'naturalisation' procedure. Glutamic acid was chosen to achieve a double glycoconjugation, which is essential to give to the original disperse dye a water solubility suitable for reaching optimal dyeing conditions. UV-vis plot of the 'naturalised' species showed negligible differences when compared to those of the commercial dyes.
Sigma-Aldrich
Disperse Red 1, Dye content 95 %