MilliporeSigma
  • Blood pressure and metabolic effect of a combination of lercanidipine with different antihypertensive drugs in clinical practice.
Blood pressure and metabolic effect of a combination of lercanidipine with different antihypertensive drugs in clinical practice.
Clinical and experimental hypertension (New York, N.Y. : 1993) (2011-10-05)
Arrigo F G Cicero, Beatrice Gerocarni, Martina Rosticci, Claudio Borghi
ABSTRACT
The aim of this study is to assess the blood pressure (BP) and metabolic effects of lercanidipine when combined with other classes of first-line antihypertensive drugs in day-to-day clinical practice. For this study, we consecutively enrolled 162 patients with uncomplicated primary hypertension, who are partial responders to the treatment with lercanidipine over a period of 24 months. Patients were then allocated to the combination of lercanidipine (10-20 mg/day) with β-blockers, diuretics, angiotensin-converting enzyme inhibitors, and angiotensin-II receptor blockers according to compelling indications (if any) and/or suggestions of European Society of Hypertension-European Society of Cardiology (ESH-ESC) guidelines. All the enrolled patients completed the study and no adverse drug reaction was registered during the research period. The association of a second drug with lercanidipine determined an additional BP decrease of either systolic BP or diastolic BP independently from the type of drug added (P always <.05). The additional effect of lercanidipine appears widely distributed with no significant differences in the size of BP decrease. From the metabolic point of view, the addition of a second drug did not determine a significant variation in the serum levels of total cholesterol, low-density lipoprotein cholesterol, and high-density lipoprotein cholesterol (P always >.05). Conversely, a significant decrease in fasting plasma glucose and serum levels of triglycerides has been observed in patients where lercanidipine has been combined with an angiotensin-converting enzyme inhibitor or an angiotensin-II receptor blocker. In conclusion, in our study we observed that lercanidipine-based protocols are well tolerated and efficacious in reducing BP. Moreover, the association of lercanidipine with renin-angiotensin system blockers is also associated with significant improvements in triglycerides and fasting plasma glucose.
Sigma-Aldrich
Lercanidipine hydrochloride, ≥98% (HPLC)