MilliporeSigma
  • Polycondensation of butenediol: synthesis of telechelic 2-butene-1,4-diol oligomers.
Polycondensation of butenediol: synthesis of telechelic 2-butene-1,4-diol oligomers.
Journal of the American Chemical Society (2011-09-24)
Matthew K Kiesewetter, Justin A Edward, Hyunuk Kim, Robert M Waymouth
ABSTRACT
The catalytic condensation of cis-2-butene-1,4-diol with CpRu(MQA)(C(3)H(5)) (Cp = cyclopentadienyl, MQA = 4-methoxyquinoline-2-carboxylate) generates poly(2-butenediol), an unsaturated telechelic polyether diol with molecular weights between 400 and 4600 g/mol. This Ru(IV) allyl catalyst enchains 2-butene-1,4-diol primarily as the linear trans-2-butenyl ether (92%) along with vinyl branches (8%). These telechelic oligomers are useful chain extenders and macromonomers, as demonstrated by their use in the synthesis of poly(lactide)-b-poly(butenediol)-b-poly(lactide) triblock copolymers. Model studies support a proposed mechanism involving the formation of Ru(IV) allyl intermediates from allylic alcohols and chain growth by selective nucleophilic displacement at the terminus of the Ru(IV) allyl to generate trans-2-butenyl ether linkages.
Sigma-Aldrich
cis-2-Butene-1,4-diol, 97%