MilliporeSigma
  • Comparison of antioxidative and antifibrotic effects of α-tocopherol with those of tocotrienol-rich fraction in a rat model of chronic pancreatitis.
Comparison of antioxidative and antifibrotic effects of α-tocopherol with those of tocotrienol-rich fraction in a rat model of chronic pancreatitis.
Pancreas (2011-09-20)
Fei Jiang, Zhuan Liao, Liang-Hao Hu, Yi-Qi Du, Xiao-Hua Man, Jun-Jun Gu, Jun Gao, Yan-Fang Gong, Zhao-Shen Li
ABSTRACT
The α-tocopherol and tocotrienol-rich fraction (TRF) are considered effective antioxidants. This study aimed to compare the antioxidative and antifibrotic effects of α-tocopherol and TFR in dibutylin dichloride (DBTC)-induced chronic pancreatitis (CP) rats. Oral administration of α-tocopherol and TFR (both 800 mg/kg per day) started the next day after DBTC (8 mg/kg) infusion into the tail vein for 4 weeks. Histological examination, Sirius red staining, and measurement of the contents of hydroxyproline and malondialdehyde of the pancreas were performed to evaluate pancreatic damage and fibrosis. Immunohistochemical analysis of α-smooth muscle actin and real-time reverse transcription polymerase chain reaction for transforming growth factor-β1 (TGF-β1) and collagen-α1(I) were performed to evaluate the activation of pancreatic stellate cells and the mRNA levels of fibrosis-related genes, respectively. Both α-tocopherol and TRF reduced oxidative stress, ameliorated inflammation and fibrosis, and down-regulated the mRNA expression of TGF-β1 and collagen-α1(I) in DBTC-induced CP. The TRF was superior to α-tocopherol in alleviating inflammation and fibrosis and down-regulating TGF-β1 mRNA expression. Oral administration of α-tocopherol and TRF improves pancreatic inflammation and fibrosis in DBTC-induced CP rats, with TRF being more effective than α-tocopherol. Therefore, TRF may be a novel option for alleviating inflammation and, particularly, the fibrotic process in CP.
Sigma-Aldrich
Dibutyltin dichloride, 96%