MilliporeSigma
  • Non-steroidal antiinflammatory drug-copper(II) complexes: structure and biological perspectives.
Non-steroidal antiinflammatory drug-copper(II) complexes: structure and biological perspectives.
Dalton transactions (Cambridge, England : 2003) (2011-08-02)
Filitsa Dimiza, Stella Fountoulaki, Athanasios N Papadopoulos, Christos A Kontogiorgis, Vassilis Tangoulis, Catherine P Raptopoulou, Vassilis Psycharis, Aris Terzis, Dimitris P Kessissoglou, George Psomas
ABSTRACT
Copper(II) complexes with the non-steroidal antiinflammatory drug mefenamic acid in the presence of aqua or nitrogen donor heterocyclic ligands (2,2'-bipyridine, 1,10-phenanthroline, 2,2'-bipyridylamine or pyridine) have been synthesized and characterized. The crystal structures of [(2,2'-bipyridine)bis(mefenamato)copper(II)], 2, [(2,2'-bipyridylamine)bis(mefenamato)copper(II)], 4, and [bis(pyridine)bis(methanol)bis(mefenamato)copper(II)], 5, have been determined by X-ray crystallography. UV study of the interaction of the complexes with calf-thymus DNA (CT DNA) has shown that the complexes can bind to CT DNA and [bis(aqua)tetrakis(mefenamato)dicopper(II)] exhibits the highest binding constant to CT DNA. The cyclic voltammograms of the complexes in the presence of CT DNA solution have shown that the complexes can bind to CT DNA by the intercalative binding mode verified also by DNA solution viscosity measurements. Competitive studies with ethidium bromide (EB) indicate that the complexes can displace the DNA-bound EB suggesting strong competition with EB. Mefenamic acid and its complexes exhibit good binding propensity to human or bovine serum albumin protein having relatively high binding constant values. All the compounds have been tested for their antioxidant and free radical scavenging activity as well as for their in vitro inhibitory activity against soybean lipoxygenase showing significant activity.
Sigma-Aldrich
Mefenamic acid