MilliporeSigma
  • Improvements in monitoring the N-terminal valine adduct in human globin after exposure to sulfur mustard and synthesis of reference chemicals.
Improvements in monitoring the N-terminal valine adduct in human globin after exposure to sulfur mustard and synthesis of reference chemicals.
Talanta (2011-07-06)
Zhiyong Nie, Qin Liu, Jianwei Xie
ABSTRACT
The N-terminal valine adduct (HETE-Val) in globin is believed to behave as a long-lived biomarker after exposure to sulfur mustard (HD). Development of a highly sensitive method for monitoring HETE-Val, particularly at low HD exposure levels or for retrospective detection, would be a significant achievement. In this study, by improving the sample preparation method, a sensitive NCI-GC/MS method was established for the analysis of HETE-Val in globin after HD exposure. To optimize and investigate the sample preparation method, all the relevant HETE-Val chemicals were synthesized, purified, and characterized. By carrying out optimized solid phase extraction (SPE) cleanup followed by modified Edman degradation results in a low detection level and clean baseline. The minimum detectable exposure level of human blood (in vitro) to HD is 20 nmol/L (S/N>3). The interday and intraday precisions of the proposed method were found to be acceptable with less than a 15% relative standard deviation (RSD). A nearly linear dose-effect relationship was observed between HETE-Val and a HD exposure concentration range of 0.1-120 μmol/L. The percentage of HD that reacted with N-terminal valine in globin obtained from human blood (in vitro) was quantified using the proposed method.
Sigma-Aldrich
N-Heptafluorobutyrylimidazole, 97%
Pricing and availability is not currently available.
Sigma-Aldrich
1-(Heptafluorobutyryl)imidazole, BioReagent, suitable for derivatization
Pricing and availability is not currently available.