MilliporeSigma
  • Harpagoside variation is positively correlated with temperature in Scrophularia ningpoensis Hemsl.
Harpagoside variation is positively correlated with temperature in Scrophularia ningpoensis Hemsl.
Journal of agricultural and food chemistry (2011-02-17)
Shuting Yang, Jinghui Li, Yunpeng Zhao, Binlong Chen, Chengxin Fu
ABSTRACT
Scrophularia ningpoensis Hemsl. is an important Chinese medicinal herb with a history of domestication of over 1000 years. Phytochemical variation of S. ningpoensis in response to environmental gradients remains an attractive topic with both practiceal and theoretical significances. In the current study, HPLC fingerprinting and four major bioactive compounds of S. ningpoensis, that is, harpagoside, angroside C, acteoside, and cinnamic acid, were determined to explore its correlations with climatic, geographic, and soil factors. The present data confirmed the approximate three-group pattern of phytochemical differentiation among the five production regions, the population of Zhejiang (ZJ), the population of Hubei (HB), and the rest three populations of Chongqing, Hunan, and Shaanxi (CQ, HN, and SX). Harpagoside, the dominant bioactive compound of S. ningpoensis, contributed most to the phytochemical differentiation and displayed a significant positive correlation with monthly and annual average temperature and negative correlations with altitude and latitude. It was concluded that harpagoside variation was strongly positively correlated with environmental changes of temperature.
Product Description
Supelco
Harpagoside, analytical standard
Harpagoside, primary reference standard