MilliporeSigma
  • Brønsted acid-catalyzed efficient Strecker reaction of ketones, amines and trimethylsilyl cyanide.
Brønsted acid-catalyzed efficient Strecker reaction of ketones, amines and trimethylsilyl cyanide.
Organic & biomolecular chemistry (2010-03-06)
Guang-Wu Zhang, Dong-Hua Zheng, Jing Nie, Teng Wang, Jun-An Ma
ABSTRACT
A general method for the one-pot, three-component Strecker reaction of ketones was developed using Brønsted acids as organocatalysts. A series of alpha-aminonitriles were obtained in good to excellent yields (79-99%). A preliminary extension to a catalytic enantioselective three-component Strecker reaction of ketones (up to 40% ee) is also described.
Sigma-Aldrich
Trimethylsilyl cyanide, technical, ≥95% (GC)
Sigma-Aldrich
Trimethylsilyl cyanide, 98%