MilliporeSigma
  • An in vivo microdialysis measurement of harpagoside in rat blood and bile for predicting hepatobiliary excretion and its interaction with cyclosporin A and verapamil.
An in vivo microdialysis measurement of harpagoside in rat blood and bile for predicting hepatobiliary excretion and its interaction with cyclosporin A and verapamil.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2009-02-25)
Qian Wu, Xiao-Dong Wen, Lian-Wen Qi, Wei Wang, Ling Yi, Zhi-Ming Bi, Ping Li
ABSTRACT
Harpagoside, a major bioactive iridoid glucoside in genus Scrophularia, has been widely used in clinical practice for the treatment of pain in the joints and lower back for its neuroprotective and anti-inflammation activities. To investigate the pharmacokinetics and hepatobiliary excretion, an in vivo microdialysis method coupled with high performance liquid chromatography was developed to monitor the concentration of harpagoside in blood and bile. The harpagoside bile-to-blood distribution ratio (AUC(bile)/AUC(blood)) up to 986.28+/-78.46 significantly decreased to 6.41+/-0.56 or 221.20+/-18.92 after co-administration of cyclosporin A or verapamil. The results indicated that harpagoside went through concentrative elimination from the bile which was probably regulated by P-glucoprotein, providing possible clinical trials of co-administration of transporter inhibitors to decrease drug efflux, thus to enhance the curative effects.
Product Description
Harpagoside, primary reference standard
Supelco
Harpagoside, analytical standard