MilliporeSigma
  • Molecular structure and catechol oxidase activity of a new copper(I) complex with sterically crowded monodentate N-donor ligand.
Molecular structure and catechol oxidase activity of a new copper(I) complex with sterically crowded monodentate N-donor ligand.
Journal of inorganic biochemistry (2009-01-13)
Adám Kupán, József Kaizer, Gábor Speier, Michel Giorgi, Marius Réglier, Ferenc Pollreisz
ABSTRACT
The attempted alkylation of 1,3-bis(2'-pyridylimino)isoindoline (indH) by the use of n-BuLi and subsequent alkyl halides led to quaternization of the pyridine nitrogens and the zwitterionic monodentate N-ligand (Me(2)ind)I was formed. By the use of the ligand the copper(I) complex [Cu(I)(Me(2)ind)I(2)] was prepared and its structure determined. It was found to be good catalyst for the oxidation of 3,5-di-tert-butylcatechol (DTBCH(2)) to 3,5-di-tert-butyl-1,2-benzoquinone (DTBQ) and H(2)O(2) by dioxygen. Detailed kinetic studies revealed first-order dependence on the catalyst and dioxygen concentration and saturation type behavior with respect to the substrate.
Sigma-Aldrich
3,5-Di-tert-butyl-o-benzoquinone, 98%