MilliporeSigma
  • Measuring the force of interaction between a metallic probe and a single molecule.
Measuring the force of interaction between a metallic probe and a single molecule.
Physical review letters (2006-10-10)
B Naydenov, P Ryan, L C Teague, John J Boland
ABSTRACT
Precision current measurements are recorded at 5 K during the approach and contact between a Pt-inked probe and the carbon-carbon double-bond region of an isolated 1,3-cyclohexadiene molecule chemisorbed on a Si(100) surface. Scanning tunneling spectroscopic data reveal systematic features in the current at specific probe-molecule separations. Aided by density functional theory calculations, we show that these features arise from interaction forces between the probe and molecule, which can be interpreted as the relaxation of the probe-molecule system prior to and during contact.
Sigma-Aldrich
1,3-Cyclohexadiene, contains 0.05% BHT as inhibitor, 97%