MilliporeSigma
  • Development and single-laboratory validation of a new gas chromatographic multi-pesticide method of analysis of commercial emulsifiable concentrate formulations containing alachlor, chlorpyrifos methyl, fenthion and trifluralin as active ingredients.
Development and single-laboratory validation of a new gas chromatographic multi-pesticide method of analysis of commercial emulsifiable concentrate formulations containing alachlor, chlorpyrifos methyl, fenthion and trifluralin as active ingredients.
Journal of chromatography. A (2006-09-06)
Helen Karasali, George Balayannis, Adamantia Hourdakis, Arpad Ambrus
ABSTRACT
A multi-pesticide (MP) method was developed and single-laboratory validated for the quality control of commercial pesticide products containing alachlor, chlorpyrifos methyl, fenthion and trifluralin as active ingredients (a.i.). A capillary gas chromatographic system with flame ionization detection (FID) and a programmable temperature vaporising split injector was used. The performance characteristics (specificity, linearity, precision and repeatability) of the method satisfied international acceptability criteria.