MilliporeSigma
  • Residue levels of captan and trichlorfon in field-treated kaki fruits, individual versus composite samples, and after household processing.
Residue levels of captan and trichlorfon in field-treated kaki fruits, individual versus composite samples, and after household processing.
Food additives and contaminants (2006-06-13)
M L Fernández-Cruz, S Grimalt, M Villarroya, F J López, S Llanos, J M García-Baudín
ABSTRACT
The dissipation of residue levels of captan and trichlorfon in field-treated kaki crops was studied according to good laboratory practices to propose maximum residue limits (MRLs). Residue levels of captan and trichlorfon were analysed by GC/MS and LC-MS/MS, respectively. Residue levels of captan and trichlorfon permitted one to propose MRLs in kaki of 3 and 5 mg kg(-1), respectively. The behaviour of these residues was also studied after peeling and cooking, and in individual fruits versus composite samples. Residue levels of these compounds for individual fruits suggested that a variability factor up to three could be set for the acute risk assessment. Levels of captan decreased by more than 90% after peeling and completely after cooking. Trichlorfon penetrates into the flesh in a proportion of 70% of the residue at the pre-harvest interval. Cooking resulted in a decrease of 27% of residue levels of trichlorfon.
Product Description
Supelco
Folpet, PESTANAL®, analytical standard