MilliporeSigma
  • Direct observation of an acyl nitroso species in solution by time-resolved IR spectrocopy.
Direct observation of an acyl nitroso species in solution by time-resolved IR spectrocopy.
Journal of the American Chemical Society (2003-02-06)
Andrew D Cohen, Bu-Bing Zeng, S Bruce King, John P Toscano
ABSTRACT
Benzoyl nitroside (5) was generated in solution by laser photolysis of 3,5-diphenyl-1,2,4-oxadiazole-4-oxide (4) and studied by time-resolved infrared spectroscopy. The second-order rate constants for reaction of 5 with diethylamine and 1,3-cyclohexadiene were determined to be (1.3 +/- 0.5) x 10(5) M(-1) s(-1) and (6.0 +/- 0.5) x 10(3) M(-1) s(-1), respectively. The formation of nitroxyl (HNO), a product of the reaction of 5 with diethylamine, was also observed.
Sigma-Aldrich
1,3-Cyclohexadiene, contains 0.05% BHT as inhibitor, 97%