MilliporeSigma
  • Carboxylic acids and skeletal muscle chloride channel conductance: effects on the biological activity induced by the introduction of an aryloxyalkyl group alpha to the carboxylic function of 4-chloro-phenoxyacetic acid.
Carboxylic acids and skeletal muscle chloride channel conductance: effects on the biological activity induced by the introduction of an aryloxyalkyl group alpha to the carboxylic function of 4-chloro-phenoxyacetic acid.
Farmaco (Societa chimica italiana : 1989) (2001-11-23)
G Carbonara, G Fracchiolla, F Loiodice, P Tortorella, D Conte-Camerino, A De Luca, A Liantonio
ABSTRACT
2-(4-Chloro-phenoxy)propanoic and 2-(4-chloro-phenoxy)butanoic acids are compounds known to block chloride membrane conductance in rat striated muscle by interaction with a specific receptor. In the present study, a series of chiral analogues has been prepared and tested to evaluate the influence of a second aryloxy moiety introduced in the side-chain at a variable distance from the stereogenic centre. The results show that this chemical modification is detrimental for biological activity which, however, is increased by lengthening the alkyl chain up to three methylenic groups, then decreases to remain constant in the next analogues of the series. A possible explanation for this is proposed on the basis of steric effects and/or different approach of the molecules to the receptor.
Product Description
Supelco
4-Chlorophenoxyacetic acid, PESTANAL®, analytical standard
Sigma-Aldrich
4-Chlorophenoxyacetic acid, BioReagent, suitable for plant cell culture, crystalline
Sigma-Aldrich
4-Chlorophenoxyacetic acid, ≥98.0% (T)