MilliporeSigma
  • A model of the quaternary structure of enolases, based on structural and evolutionary analysis of the octameric enolase from Bacillus subtilis.
A model of the quaternary structure of enolases, based on structural and evolutionary analysis of the octameric enolase from Bacillus subtilis.
Journal of protein chemistry (1999-02-13)
C K Brown, P L Kuhlman, S Mattingly, K Slates, P J Calie, W W Farrar
ABSTRACT
Purified enolase from Bacillus subtilis has a native mass of approximately 370 kDa. Since B. subtilis enolase was found to have a subunit mass of 46.58 kDa, the quaternary structure of B. subtilis is octameric. The pl for B. subtilis enolase is 6.1, the pH optimum (pHo) for activity is 8.1-8.2, and the Km for 2-PGA is approximately 0.67 mM. Using the dimeric Calpha structure of yeast dimeric enolase as a guide, these dimers were arranged as a tetramer of dimers to simulate the electron microscopy image processing obtained for the octameric enolase purified from Thermotoga maritima. This arrangement allowed identification of helix J of one dimer (residues 86-96) and the loop between helix L and strand 1 (HL-S1 loop) of another dimer as possible subunit interaction regions. Alignment of available enolase amino acid sequences revealed that in 16 there are two tandem glycines at the C-terminal end of helix L and the HL-S1 loop is truncated by 4-6 residues relative to the yeast polypeptide, two structural features absent in enolases known to be dimers. From these arrangements and alignments it is proposed that the GG tandem at the C-terminal end of helix L and truncation of the HL-S1 loop may play a critical role in octamer formation of enolases. Interestingly, the sequence features associated with dimeric quaternary structure are found in three phylogenetically disparate groups, suggesting that the ancestral enolase was an octamer and that the dimeric structure has arisen independently multiple times through evolutionary history.
Sigma-Aldrich
L-2-Phosphoglyceric acid disodium salt hydrate, ≥80% (CE)