MilliporeSigma
  • Rapid synthesis of a 5'-fluorinated oligodeoxy-nucleotide: a model antisense probe for use in imaging with positron emission tomography (PET).
Rapid synthesis of a 5'-fluorinated oligodeoxy-nucleotide: a model antisense probe for use in imaging with positron emission tomography (PET).
Bioorganic & medicinal chemistry letters (1999-01-01)
D Pan, S S Gambhir, T Toyokuni, M R Iyer, N Acharya, M E Phelps, J R Barrio
ABSTRACT
5'-Deoxy-5'-fluoro-O4-methylthymidine was synthesized by the reaction of the corresponding 5'-O-tosylate with KF in the presence of Kryptofix [222] and coupled to a 5'-phosphoramidite-activated CPG-bound oligodeoxynucleotide. The sequence of reactions and purifications were accomplished within 4 h, a necessary condition of the development of radiofluorinated antisense oligodeoxynucleotide probe for use with PET.
Sigma-Aldrich
4,7,13,16,21,24-Hexaoxa-1,10-diazabicyclo[8.8.8]hexacosane, 98%