Merck
  • Simple method for the determination of 5-aminosalicylic and N-acetyl-5-aminosalicylic acid in rectal tissue biopsies.
Simple method for the determination of 5-aminosalicylic and N-acetyl-5-aminosalicylic acid in rectal tissue biopsies.
Journal of chromatography. B, Biomedical sciences and applications (1998-11-21)
F N Hussain, R A Ajjan, M Moustafa, J C Anderson, S A Riley
ABSTRACT
We describe a sensitive high-performance liquid chromatographic method for the determination of 5-aminosalicylic acid and N-acetyl-5-aminosalicylic acid in rectal tissue biopsies. Samples were derivatised using propionic anhydride and proteins were precipitated with methanol. A Supelcosil ABZ column (150x4.6 mm I.D., 5 microm silica particles) was used with a mobile phase comprising 0.1 M acetic acid, acetonitrile and triethylamine (1600:114:6, v/v/v). Fluorescence detection was employed and detection limits were 0.2 ng/mg tissue at a signal-to-noise ratio of three (measured concentration: 5-aminosalicylic acid, 0.254 (0.228-0.286) ng/mg, C.V. 10.7%; N-acetyl-5-aminosalicylic acid, 0.18 (0.154-0.198) ng/mg, C.V 9.8%). This assay was validated for use with serum, urine and faecal samples for which it proved to be both precise and accurate (C.V.<10%, measured concentration within 10%).
Sigma-Aldrich
Propionic anhydride, purum, ≥96.0% (NT)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Propionic anhydride, 97%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Propionic anhydride, ≥99%
SKU
Pack Size
Availability
Price
Quantity
Supelco
SUPELCOSIL LC-NH2-NP HPLC Column, 5 μm particle size, L × I.D. 25 cm × 4.6 mm
SKU
Pack Size
Availability
Price
Quantity
Supelco
SUPELCOSIL LC-NH2-NP Supelguard Cartridge, 5 μm particle size, L × I.D. 2 cm × 4 mm