MilliporeSigma
  • Modulation of proto-oncogene expression by polychlorinated biphenyls in 3T3-L1 cell line.
Modulation of proto-oncogene expression by polychlorinated biphenyls in 3T3-L1 cell line.
Journal of toxicology and environmental health. Part A (1998-10-07)
L Gribaldo, M G Sacco, S Casati, I Zucchi, M K Dosanjh, P Catalani, E Marafante
ABSTRACT
The effects of two substituted polychlorinated biphenyls, the 3,4,5,3',4,5' (PCB-169) and the 2,3,4,2',4',5' (PCB-138) forms, were examined on the expression of c-myc, c-jun, c-ras, and jun-b in 3T3-L1 cells. Northern blot analysis demonstrated that the two PCBs, which exhibit a coplanar and di-ortho-substituted configuration, activated these oncogenes differently. PCB-138 markedly induced overexpression of ras, jun, and myc, whereas PCB-169 led to the overexpression of jun-b. High-performance liquid chromatography analysis of the cell samples treated in medium without serum revealed a higher intracellular concentration of the 2,3,4,2',4',5'-hexachlorobiphenyl (hexaCB), whereas the 3,4,5,3',4'5'-hexaCB reached the same concentration in the sonicated samples of cells with or without serum. These results indicated that there was a relationship between PCB structure, bioavailability, and the capacity to stimulate oncogene expression.
Product Description
Supelco
PCB No 138, analytical standard
2,2′,3,4,4′,5′-Hexachlorobiphenyl (IUPAC No. 138), BCR®, certified reference material