MilliporeSigma
  • Characterization of acetylcholinesterase purified from the lesser grain borer, Rhyzopertha dominica (Coleoptera: Bostrichidae).
Characterization of acetylcholinesterase purified from the lesser grain borer, Rhyzopertha dominica (Coleoptera: Bostrichidae).
Comparative biochemistry and physiology. Part C, Pharmacology, toxicology & endocrinology (1998-07-21)
R N Guedes, K Y Zhu, S Kambhampati, B A Dover
ABSTRACT
Acetylcholinesterase (AChE, EC 3.1.1.7) purified from the lesser grain borer (Rhyzopertha dominica) was significantly inhibited by higher concentrations of the substrates acetylthiocholine (ATC), acetyl-(beta-methyl) thiocholine (A beta MTC) and propionylthiocholine (PTC). 2. The efficiency of AChE for hydrolyzing different substrates was ATC > A beta MTC > PTC > S-butyrylthiocholine. The enzyme activity was completely inhibited by 10(-5) M eserine or BW284C51, but was only partially inhibited by ethopropazine at the same concentration. These results confirmed that the purified enzyme was an typical insect AChE. 3. Non-denaturing and SDS polyacrylamide gel electrophoresis (PAGE) showed only one major molecular form in the purified AChE with a molecular weight of about 107,000 prior to reduction and about 56,000 after reduction, suggesting the homodimer of AChE linked with disulfide bonds.
Sigma-Aldrich
Propionylthiocholine iodide, ≥98%