MilliporeSigma
  • Effect of selective substitution of 5-bromocytosine on conformation of DNA triple helices.
Effect of selective substitution of 5-bromocytosine on conformation of DNA triple helices.
Journal of biomolecular structure & dynamics (1998-06-10)
M Liu, L Yang, W Deng, M Su, C Wang, S B Lin, L S Kan, C Bai
ABSTRACT
Three triplex DNAs containing 5-bromocytosine[BrC] were studied by vibrational spectroscopy and molecular modelling. Firstly, three oligodeoxypyrimidines of 5'-(TC)3-T4-(BrCT)3 [CBrC], 5'-(TBrC)3-T4-(CT)3 [BrCC] and 5'-(TBrC)3-T4-(BrCT)3 [BrCBrC] were synthesized and then reacted with an oligodeoxypurine of 5'-(AG)3 at pH=4.5 in phosphate buffer respectively to form three comparative hairpin triplex named CY,YC and YY. The results of FT-Raman and IR revealed that YY is almost in A-like form, CY and YC are combinations of A-like form and B-like form, but A-form dominates in CY while B-form is equivalent as A-form in YC. The result is consistent with the theoretical analysis.