MilliporeSigma
  • Cardiac sensitization testing of the halon replacement candidates trifluoroiodomethane (CF3I) and 1,1,2,2,3,3,3-heptafluoro-1-iodopropane (C3F7I).
Cardiac sensitization testing of the halon replacement candidates trifluoroiodomethane (CF3I) and 1,1,2,2,3,3,3-heptafluoro-1-iodopropane (C3F7I).
Drug and chemical toxicology (1998-05-23)
D E Dodd, A Vinegar
ABSTRACT
Trifluoroiodomethane (CF3I) and 1,1,2,2,3,3,3-heptafluoro-1-iodopropane (C3F7I) have been considered as replacement candidates for halon fire suppressants due to their excellent fire extinguishant capabilities and low ozone depletion potential compared to halon fire extinguishants in use currently. As part of the process to develop environmental and health effects criteria for halon substitutes, a cardiac sensitization test was conducted in beagle dogs. Cardiac sensitization to adrenaline is a phenomenon associated with the inhalation of a number of unsubstituted and halogenated hydrocarbons. Adrenaline was administered by intravenous injection before and during inhalation of the test substance. CF3I was administered to dogs at concentrations in air of 0.1, 0.2, 0.4 or 1% v/v. At each of 0.4 and 1.0% CF3I, the first dog exposed developed fatal ventricular fibrillation, and no further dogs were exposed at these concentrations. There was no cardiac sensitization at 0.1 or 0.2% CF3I. For the C3F7I experiment, dogs were exposed to concentrations in air of 0.1, 0.2 or 0.4% v/v. At each of 0.1 and 0.4% C3F7I, one dog responded with multifocal ventricular ectopic beats. Thus, CF3I and C3F7I are potent cardiac sensitizers in the adrenaline-challenged dog model.
Sigma-Aldrich
Perfluoropropyl iodide, 98%