MilliporeSigma
Labeling of penicillamine di sulfide with technetium-99m.
Applied radiation and isotopes : including data, instrumentation and methods for use in agriculture, industry and medicine (1998-05-07)
P Unak, M Tunç, Y Duman
ABSTRACT
Complex forming conditions of Penicillamine di sulfide with 99mTc have been specified. Labeling of penicillamine di sulfide with 99mTc by direct reduction with SnCl2 did not give favorable good results while the 99mTc complex of penicillamine can be easily obtained. Ligand exchange reaction with 99mTc-gluconate was attempted and a 95% labeling efficiency was obtained. Radiopharmaceutical potential of 99mTc-PADS (99mTc-Penicillamine di sulfide) has been investigated with a gamma camera in rabbits and the complex was found to be uptaken mostly by the liver and kidneys.
Sigma-Aldrich
D-Penicillamine disulfide, 97%