MilliporeSigma
  • Absorbable suture technique: solution to the growth problem in pediatric pacing with endocardial leads.
Absorbable suture technique: solution to the growth problem in pediatric pacing with endocardial leads.
Pacing and clinical electrophysiology : PACE (1998-02-25)
P Stojanov, D Velimirović, V Hrnjak, S U Pavlović, M Zivković, Z Djordjević
ABSTRACT
Endocardial pacing system implantation has been performed in 15 children of mean age 37 months (ranging from 1 day to 89 months). Endocardial lead fixation was performed by means of slowly resorbable suture (Dexon) to allow spontaneous lead migration as the child grows. During a mean follow-up period of 61 months (range 17-108 months), none of the patients needed reintervention for correcting the lead length to allow growth.
Sigma-Aldrich
Fenaminosulf, ≥98%
Supelco
Fenaminosulf, PESTANAL®, analytical standard