MilliporeSigma
  • Modification of IgH PCR clonal analysis by the addition of sucrose and cresol red directly to PCR reaction mixes.
Modification of IgH PCR clonal analysis by the addition of sucrose and cresol red directly to PCR reaction mixes.
Molecular pathology : MP (1997-06-01)
E Hodges, S M Boddy, S Thomas, J L Smith
ABSTRACT
Diagnostic immunoglobulin (Ig) polymerase chain reaction (PCR) clonality analyses need to be simple, reproducible, and rapid. Sucrose and cresol red (gel loading buffer reagents) were added to a routine IgH PCR reaction mix to obviate the need for adding gel loading buffer separately after PCR amplification. Not only did this decrease the time spent after PCR analysis but also gave similar or enhanced IgH PCR product intensity compared with normal IgH PCR conditions on polyacrylamide gel electrophoresis. This procedure was easily adapted to routine PCR analysis without the need for further manipulations or optimisation of the PCR reaction mix, and it increased the reproducibility and specificity of the IgH PCR products.
Sigma-Aldrich
Cresol Red sodium salt, indicator grade
Sigma-Aldrich
Cresol red, indicator grade, Dye content 95 %