MilliporeSigma
Evolution of bacterial genomes.
Antonie van Leeuwenhoek (1997-03-01)
J T Trevors
ABSTRACT
This review examines evolution of bacterial genomes with an emphasis on RNA based life, the transition to functional DNA and small evolving genomes (possible plasmids) that led to larger, functional bacterial genomes.
Sigma-Aldrich
3,5-Diamino-1,2,4-triazole, 98%