MilliporeSigma
  • Determination of L-carnitine, acetyl-L-carnitine and propionyl-L-carnitine in human plasma by high-performance liquid chromatography after pre-column derivatization with 1-aminoanthracene.
Determination of L-carnitine, acetyl-L-carnitine and propionyl-L-carnitine in human plasma by high-performance liquid chromatography after pre-column derivatization with 1-aminoanthracene.
Journal of chromatography. B, Biomedical applications (1996-11-15)
A Longo, G Bruno, S Curti, A Mancinelli, G Miotto
ABSTRACT
A new sensitive high-performance liquid chromatographic procedure for the determination of L-carnitine (LC), acetyl-L-carnitine (ALC) and propionyl-L-carnitine (PLC) in human plasma has been developed. Precolumn derivatization with 1-aminoanthracene (1AA), performed in phosphate buffer in the presence of 1-(3-dimethylaminopropyl)-3-ethylcarbodiimide hydrochloride (EDC) as catalyst, is involved. The fluorescent derivatives were isocratically separated on a reversed-phase column (C18). The eluate was monitored with a fluorimetric detector set at 248 nm (excitation wavelength) and 418 nm (emission wavelength). Because of the presence of endogenous carnitines, the validation was performed using dialyzed plasma. The identity of the derivatized compounds was assessed by mass spectrometry and the purity of the chromatographic peaks was confirmed by HPLC-tandem mass spectrometry. The limits of quantitation were 5 nmol/ml for LC, 1 nmol/ml for ALC and 0.25 nmol/ml for PLC. The recovery of the extraction procedure was in the range 82.6%-95.4% for all 3 compounds. Good linearity (R approximately 0.99) was observed within the calibration ranges studied: 5-160 nmol/ml for LC, 1-32 nmol/ml for ALC and 0.25-8 nmol/ml for PLC. Precision was in the range 0.3-16.8% and accuracy was always lower than 10.6%.
Sigma-Aldrich
1-Aminoanthracene, technical grade, 90%