MilliporeSigma
  • Simultaneous determination of phenol, cresol, xylenol isomers and naphthols in urine by capillary gas chromatography.
Simultaneous determination of phenol, cresol, xylenol isomers and naphthols in urine by capillary gas chromatography.
Journal of chromatography. B, Biomedical applications (1996-06-28)
G Bieniek
ABSTRACT
An attempt was made to establish a method for the simultaneous determination of urinary concentrations of phenol, o-, p- and m-cresols, 1- and 2-naphthol and xylenol isomers by capillary gas chromatography. Urine samples were extracted after acid hydrolysis of glucuronides and sulfates by solid-phase extraction. The ten substances were separated gas chromatographically using a capillary column (Ultra 2) of cross-linked 5% phenylmethyl silicone. Calibration graphs were linear for 5-100 micrograms/ml of all the phenols determined. The corresponding detection limits for phenolic compounds varied from 0.1 to 0.2 microgram/ml. The relative standard deviations for samples in urine were in the range 2.6 - 16.6% and the accuracy was in the range 1.4-25%. Recoveries were generally over 80%.
Sigma-Aldrich
3,5-Dimethylphenol, 98%
Sigma-Aldrich
3,5-Dimethylphenol, ≥99%