MilliporeSigma
  • Fragmentation of isomeric N-quinolinylphthalimides on electron impact ionization.
Fragmentation of isomeric N-quinolinylphthalimides on electron impact ionization.
Journal of mass spectrometry : JMS (1996-06-01)
A Weisz, D Andrzejewski, A Mandelbaum
ABSTRACT
The isomeric 2-, 3-, 5-, 6- and 8-quinolinylphthalimides give rise to different electron impact ionization mass spectra, which permit easy distinction. The specific fragmentation process are rationalized in terms of proximity effects and stabilization of cyclic ion structures. Collision-induced dissociation spectra were used to support the proposed ion structures of major fragment ions.
Sigma-Aldrich
Quinoline Yellow, for microscopy (Hist.), mixture of mono- and disulfonic acid sodium salt
Sigma-Aldrich
Quinoline Yellow, Mixture of the mono- and disulfonic acids of Quinoline Yellow