MilliporeSigma
Contact stomatitis due to palladium and platinum in dental alloys.
Contact dermatitis (1996-04-01)
P Koch, H P Baum
ABSTRACT
We report a patient with contact stomatitis due to combined sensitization to palladium and platinum. Patch testing showed strong and persistent allergic patch test reactions to palladium chloride (1% pet.), ammonium tetrachloroplatinate (0.25% pet.), and a palladium metal plate. A platinum metal plate showed a weaker reaction. Histological examination of a biopsy from the test site of palladium chloride (1% pet.) at D3 showed both eczematous and lichenoid changes.
Sigma-Aldrich
Ammonium tetrachloroplatinate(II), 99%
Sigma-Aldrich
Potassium tetrachloroplatinate(II), 98%
Sigma-Aldrich
Potassium tetrachloroplatinate(II), 99.99% trace metals basis
Sigma-Aldrich
Potassium tetrachloroplatinate(II), ≥99.9% trace metals basis