MilliporeSigma
  • An improved method for rapid analysis of the fatty acids of glycerolipids.
An improved method for rapid analysis of the fatty acids of glycerolipids.
Lipids (1996-05-01)
K Ichihara, A Shibahara, K Yamamoto, T Nakayama
ABSTRACT
An improved rapid procedure to determine the fatty acid composition of glycerolipids is described. The procedure includes KOH-catalyzed transesterification and high-speed gas chromatography. Glycerolipids (20-40 mg) were mixed with 2 mL of hexane and 0.2 mL of 2 M methanolic KOH at room temperature for 1-2 min. The fatty acid methyl esters in the hexane layer were analyzed by gas chromatography on 10% SP-2340 at 240 degrees C. Methyl linolenate and docosahexaenoate eluted within 2 and 5 min, respectively. Analysis was thus completed within 5 min for common vegetable oils and 8 min for fish oils.
Sigma-Aldrich
Oleyl oleate, ≥99%
Supelco
SP®-2340 Capillary GC Column, L × I.D. 60 m × 0.32 mm, df 0.20 μm
Supelco
SP®-2340 Capillary GC Column, L × I.D. 30 m × 0.25 mm, df 0.20 μm
Supelco
SP®-2340 Capillary GC Column, L × I.D. 60 m × 0.25 mm, df 0.20 μm