MilliporeSigma
  • Identification of topaquinone, as illustrated for pig kidney diamine oxidase and Escherichia coli amine oxidase.
Identification of topaquinone, as illustrated for pig kidney diamine oxidase and Escherichia coli amine oxidase.
Analytical biochemistry (1995-09-01)
V Steinebach, B W Groen, S S Wijmenga, W M Niessen, J A Jongejan, J A Duine
ABSTRACT
Pig kidney diamine oxidase was purified to homogeneity. The reaction product of the cofactor with p-nitrophenylhydrazine (pNPH) was liberated with pronase treatment and purified. 1H NMR, uv/vis, and electrospray tandem mass spectroscopy revealed it to be a dipeptide with the sequence topaquinone-pNPH and aspartate. No heterogeneity was observed, indicating that no intramolecular cyclization of the quinone moiety occurs in the time span of the isolation and of the measurements. Similar results were obtained with the more widely applicable reagent, phenylhydrazine, and using the aromatic amine oxidase from Escherichia coli. From the amount and ease with which the dipeptide could be isolated, the procedure used here is more convenient than the existing one for the identification of protein-integrated quinone cofactors.