MilliporeSigma
  • Synthesis of 2- and 4-nitrophenyl beta-glycosides of beta-(1-->4)- D-xylo-oligosaccharides of dp 2-4.
Synthesis of 2- and 4-nitrophenyl beta-glycosides of beta-(1-->4)- D-xylo-oligosaccharides of dp 2-4.
Carbohydrate research (1995-11-22)
K Takeo, Y Ohguchi, R Hasegawa, S Kitamura
ABSTRACT
2- and 4-Nitrophenyl beta-D-xylopyranosides (4 and 5) were transformed, via dibutyltin oxidemediated acylation, into the corresponding 2,3-di-O-benzoyl derivatives 11 and 15. Xylobiose and xylotriose were easily isolated by charcoal column chromatography from a commercially available material and converted into the di- and trisaccharide methyl 1-thio-beta-glycosides 36 and 37. The 2-and 4-nitrophenyl beta-glycosides of the beta-(1-->4)-D-xylo-oligosaccharides of dp 2-4 were synthesized by N-iodosuccinimide-silver triflate-promoted condensation using 11 and 15 as the glycosyl acceptors and ethyl 1-thio-beta-D-xylopyranoside triacetate 16, 36, and 37 as the glycosyl donors. Also described are an improved preparation of 4 and 5, and the synthesis of 1-naphthyl beta-D-xylopyranoside, as well as an alternative approach to the 2- and 4-nitrophenyl beta-xylobiosides.
Sigma-Aldrich
Dibutyltin(IV) oxide, 98%