MilliporeSigma
  • Osmotic effectors and DNA structure: effect of glycine on precipitation of DNA by multivalent cations.
Osmotic effectors and DNA structure: effect of glycine on precipitation of DNA by multivalent cations.
Journal of biomolecular structure & dynamics (1995-08-01)
S Flock, R Labarbe, C Houssier
ABSTRACT
We have investigated the effect of glycine (an organic osmolyte) on DNA precipitation induced by spermine4+, spermidine3+ and Tb3+ addition, using circular dichroism (CD), UV spectroscopy (UV), and electric linear dichroism (ELD) techniques. DNA precipitation by the three compounds is perturbed by glycine: more spermine4+, spermidine3+ and Tb3+ must be added to obtain the same extent of precipitation as compared to the behaviour in absence of this organic osmolyte. It seems that glycine has a general effect on the DNA environment. Calculations based on experimental results and Manning's counterion condensation theory show that glycine could modify the electrostatic environment of DNA as a consequence of a change in dielectric constant.
Sigma-Aldrich
Terbium(III) chloride hexahydrate, 99.999% trace metals basis
Sigma-Aldrich
Terbium(III) chloride, anhydrous, powder, 99.9% trace metals basis
Sigma-Aldrich
Terbium(III) chloride, anhydrous, powder, 99.99% trace metals basis
Sigma-Aldrich
Terbium(III) chloride hexahydrate, 99.9% trace metals basis