MilliporeSigma
  • Analysis of regional hemodynamic regulation in response to scald injury.
Analysis of regional hemodynamic regulation in response to scald injury.
The Journal of clinical investigation (1994-01-01)
P A Taheri, H L Lippton, S D Force, E W Franklin, A L Hyman, L M Flint, J J Ferrara
ABSTRACT
Ultrasonic probes were placed around dog femoral arteries to record blood flow. Hind paw scalding with boiling water (5 s) caused a marked increase in ipsilateral femoral blood flow that persisted for the 2-h observation period. Contralateral femoral blood flow and systemic and pulmonary vascular resistances were unchanged. Compared to scald only animals, methysergide pretreatment diminished and shortened the femoral vasodilator response to scald (109 +/- 14 vs 243 +/- 27 ml/min at 5 min; 59 +/- 14 vs 191 +/- 31 ml/min at 2 h). Pretreatment with ritanserin, BW A1433U83, atropine, ICI 118551, diphenhydramine, ranitidine, meclofenamate, L-nitro-arginine methyl ester, 3-amino-1,2,4-triazine, and U 37883A had no effect on the increased femoral blood flow response to scald, suggesting this vasodilator response is not dependent upon activation of serotonergic2, adenosineA1, muscarinic, beta 2-adrenergic, histaminergic1 or histaminergic2 receptors, on cyclooxygenase products, endothelium-derived relaxing factor derived from nitric oxide (NO) synthase III, NO derived from NO synthase II, or KATP channels, respectively. Methysergide given after burn immediately reduced the augmented femoral blood flow to preburn levels, suggesting the vasodilator response to scald is mediated through continual activation of local serotonergic1-like receptors, which may be target site(s) for therapeutic interventions to influence burn-induced hemodynamic alterations.
Sigma-Aldrich
3-Amino-1,2,4-triazine, 97%