MilliporeSigma
  • Structures of bacitracin A and isolated congeners: sequencing of cyclic peptides with blocked linear side chains by electrospray ionization mass spectrometry.
Structures of bacitracin A and isolated congeners: sequencing of cyclic peptides with blocked linear side chains by electrospray ionization mass spectrometry.
Biological mass spectrometry (1994-04-01)
M M Siegel, J Huang, B Lin, R Tsao, C G Edmonds
ABSTRACT
The bacitracin antibiotic complex consists principally of bacitracin A, a peptide antibiotic containing seven amino acid residues in a ring and five amino acid residues in a blocked side chain, together with a mixture of minor components presumably related but of unknown structures. A preparative high-performance liquid chromatographic method was developed for isolating the minor components A2, B1 and B2 which were then characterized by amino acid analysis, exact mass fast atom bombardment (FAB) mass spectrometry, FAB tandem mass spectrometry (MS/MS) and electrospray ionization (ESI) mass spectrometry. For bacitracins A (MW 1421), A2 (MW 1421), B1a (MW 1407), B1b (MW 1407), B2 (MW 1407) and F (MW 1419), the side chain sequences were determined by ESI MS/MS and ESI nozzle-skimmer collision-induced dissociation (CID) mass spectrometry and the ring sequences elucidated by ESI nozzle-skimmer CID MS/MS. Relative to bacitracin A, bacitracin A2a has the modified isoleucine residue at position 1 replaced by a modified allo-isoleucine residue, bacitracin B1a has the isoleucine residue at position 8 replaced by a valine residue, bacitracin B1b has the isoleucine residue at position 5 replaced by a valine residue and bacitracin B2 has the modified isoleucine residue at position 1 replaced by a modified valine residue. FAB tandem mass spectra were shown to be consistent with the above structural assignments for the isolated bacitracin components. Structures were also proposed for the trace bacitracin components C1 (MW 1393) and D1 (MW 1379) using ESI MS/MS data obtained from the analysis of the bacitracin complex without isolation.
Product Description
Supelco
Bacitracin A, VETRANAL®, analytical standard, main stereoisomer ~50 %
Pricing and availability is not currently available.