MilliporeSigma
  • Fluorometric study of the molecular states of 2,5-diphenyloxazole in ground mixtures with gamma-cyclodextrin.
Fluorometric study of the molecular states of 2,5-diphenyloxazole in ground mixtures with gamma-cyclodextrin.
Pharmaceutical research (1994-02-01)
K Yamamoto, T Oguchi, E Yonemochi, Y Matsumura, Y Nakai
ABSTRACT
Molecular states of 2,5-diphenyloxazole (PPO) were investigated in ground mixtures with gamma-cyclodextrin (gamma-CD). Crystalline PPO gradually became amorphous upon grinding in the presence of gamma-CD. Solid-state fluorescence spectra of the ground mixtures showed that the fluorescence emission peak due to PPO crystals was reduced in intensity with an increasing duration of grinding. A new fluorescence peak attributed to PPO excimer appeared at a longer wave-length. Results of the time-resolved fluorescence study indicated that the cogrinding caused an increase in the portion of PPO excimer in the gamma-CD ground mixture. On the other hand, only a small portion of excimer was formed in ground mixtures with beta-CD, which has a smaller cavity than gamma-CD. These results suggest that cogrinding of PPO with gamma-CD caused two PPO molecules to be included in the gamma-CD cavity, contributing to excimer formation.
Sigma-Aldrich
2,5-Diphenyloxazole, suitable for liquid scintillation spectrometry
Sigma-Aldrich
2,5-Diphenyloxazole, 99%, suitable for scintillation