MilliporeSigma
  • Rapid identification of some coumarin derivatives in deterpenated citrus peel oil by gas chromatography.
Rapid identification of some coumarin derivatives in deterpenated citrus peel oil by gas chromatography.
Journal of chromatography. A (1994-06-24)
D Chouchi, D Barth
ABSTRACT
Generally on the gas chromatogram of a volatile essential oil, terpenes, oxygenated compounds and sesquiterpenes appear. With temperature programming, it was shown that some non-volatiles are present with the volatiles. They are simple coumarin (2H-1-benzopyran-2-one) derivatives such as citropten (5,7-dimethoxycoumarin) and furocoumarins (psoralen, 7H-furo[3,2-g][1]benzopyran-7-one) such as bergapten (5-methoxypsoralen), some of which are phototoxic. Terpeneless oils are used in perfumes and cosmetics, so it is important to be able to establish rapidly if they contain phototoxic compounds.
Sigma-Aldrich
5,7-Dimethoxycoumarin, 98%
Pricing and availability is not currently available.