MilliporeSigma
  • [The beneficial effects of immunostimulating and antiviral therapy on ophthalmic zona].
[The beneficial effects of immunostimulating and antiviral therapy on ophthalmic zona].
Revue roumaine de virologie (Bucharest, Romania : 1990) (1993-01-01)
V Topciu, G Chercotă, R Mihăilescu, A Nadolnic
ABSTRACT
A group of 16 patients with ophthalmic zona zoster received antiviral and immunostimulating treatment with Romanian specific products. Results were spectacular. The problem of the identity of the varicella and zoster viruses is discussed.
Sigma-Aldrich
Moroxydine hydrochloride, 99%