MilliporeSigma
Methoxatin (PQQ) in guinea-pig neutrophils.
Free radical biology & medicine (1994-10-01)
A Bishop, M A Paz, P M Gallop, M L Karnovsky
ABSTRACT
PQQ, also called methoxatin, has been isolated from guinea-pig neutrophils. The organic cations diphenyleneiodonium (DPI) and diphenyliodonium (BPI) and the aromatic o-diamine 4,5-dimethylphenylenediamine (DIMPDA) sequester synthetic PQQ and inhibit its redox-cycling activity in a model system. Standards were made of adducts of tritiated PQQ with unlabeled DIMPDA and of unlabeled PQQ with tritiated DPI or DIMPDA. PQQ adducts were isolated from guinea-pig neutrophils with each of the tritiated inhibitors. They were separated and defined by high-performance liquid chromatography (HPLC). Tiron, a disodium benzene disulphonic acid, broke the DPI-PQQ adduct isolated from neutrophils and released free PQQ. Both DPI and DIMPDA, as well as BPI, blocked O2.- release by stimulated neutrophils. The blockade exerted by these inhibitors was released by the addition of PQQ to the cell suspensions. The data demonstrate the presence of PQQ in guinea-pig neutrophils and suggest that it has a possible role, direct or indirect, in the O2.(-)-producing respiratory burst.
Sigma-Aldrich
4,5-Dimethyl-1,2-phenylenediamine, 98%