MilliporeSigma
  • Optimization by mass spectrometry of a tryptophan-specific protein cleavage reaction.
Optimization by mass spectrometry of a tryptophan-specific protein cleavage reaction.
Rapid communications in mass spectrometry : RCM (1994-09-01)
M M Vestling, M A Kelly, C Fenselau
ABSTRACT
Mass spectrometry has been used to define the reaction of BNPS-skatole (3-bromo-3-methyl-2-(o-nitrophenylsulfenyl)indolenine) with selected proteins and peptides, confirming the cleavage of the protein chain at tryptophan residues, characterizing the products, and identifying the side reactions. A short procedure for carrying out the cleavage reaction with minimal side reactions is presented.