MilliporeSigma
5-Nitroindole as an universal base analogue.
Nucleic acids research (1994-10-11)
D Loakes, D M Brown
ABSTRACT
4-, 5- and 6-Nitroindole have been investigated and compared with 3-nitropyrrole as universal bases in oligodeoxynucleotides. Of these the 5-nitroindole derivative was found to be superior giving higher duplex stability, and behaving indiscriminately towards each of the four natural bases in duplex melting experiments. 3-Nitropyrrole, whilst not discriminating between the natural bases, was found to lead to considerable destabilisation of the duplexes, particularly when multiple substitutions were made, in contrast to the 5-nitroindole nucleoside.
Sigma-Aldrich
5-Nitroindole, 98%