MilliporeSigma
Diagnostic pearls in occupational dermatology.
Dermatologic clinics (1994-07-01)
M Bruze
ABSTRACT
The author presents four cases in which investigation has led to new knowledge of causes and new diagnostic tools for occupational dermatoses.
Sigma-Aldrich
1,3-Diphenylguanidine, 97%
Supelco
1,3-Diphenylguanidine, analytical standard (for titrimetry)