MilliporeSigma
  • Micellar capillary electrophoresis separation and thermo-optical absorbance detection of products from manual peptide sequencing.
Micellar capillary electrophoresis separation and thermo-optical absorbance detection of products from manual peptide sequencing.
Electrophoresis (1994-10-01)
M Chen, K C Waldron, Y Zhao, N J Dovichi
ABSTRACT
Micellar capillary electrophoresis is optimized for separation of phenylthiohydantoin (PTH) amino acids produced in manual Edman degradation reaction for protein sequencing. There are also two major side-products produced by the Edman degradation reaction: diphenylthiourea and dimethylphenylthiourea. We report the complete separation of 19 PTH amino acids plus the two major side-reaction products in 10 min. Capillary electrophoresis is used to identify the five residues generated by manual Edman degradation sequencing of a pentapeptide.
Sigma-Aldrich
N,N′-Diphenylthiourea, 98%
Pricing and availability is not currently available.