MilliporeSigma
  • Hemorrhagic principles in the venom of Bitis arietans, a viperous snake. I. Purification and characterization.
Hemorrhagic principles in the venom of Bitis arietans, a viperous snake. I. Purification and characterization.
Biochimica et biophysica acta (1995-01-05)
T Omori-Satoh, Y Yamakawa, Y Nagaoka, D Mebs
ABSTRACT
Two hemorrhagic principles (Bitis arietans hemorrhagin a and b: abbreviated as BHRa and BHRb) were purified from the venom of the viperous snake Bitis arietans (puff adder) by gel filtration, ion-exchange and absorption chromatography. A 10-fold purification was achieved for BHRa and 7-fold for BHRb with an overall yield of 6.4% of hemorrhagic activity. The hemorrhagins were homogeneous according to disc- and SDS-polyacrylamide gel electrophoresis and immunodiffusion. BHRa and BHRb consist of 623 and 685 amino-acid residues and their apparent molecular weights were 68,000 and 75,000, respectively. They were also immunologically distinct. The purified hemorrhagins express proteolytic activity with heat-denatured casein and hide powder azure. The proteolytic activity with heat-denatured casein was almost the same as that of the crude venom, but that with hide powder azure was less than one-tenth of that of the crude venom. The purified hemorrhagins were free of arginine esterase and phospholipase A2 activities and they are acid labile hemorrhagic toxins. Their hemorrhagic activity was inhibited by EDTA, cysteine and by polyvalent anti-snake serum, but not by phenylmethanesulfonyl fluoride or soybean trypsin inhibitor.
Sigma-Aldrich
Hide–Remazol Brilliant Blue R, protease substrate