MilliporeSigma
  • Synthesis and anti-HIV activity of thiocholesteryl-coupled phosphodiester antisense oligonucleotides incorporated into immunoliposomes.
Synthesis and anti-HIV activity of thiocholesteryl-coupled phosphodiester antisense oligonucleotides incorporated into immunoliposomes.
Antiviral research (1994-09-01)
O Zelphati, E Wagner, L Leserman
ABSTRACT
Encapsulation of oligonucleotides in antibody-targeted liposomes (immunoliposomes) which bind to target cells permits intracellular delivery of the oligonucleotides. This approach circumvents problems of extracellular degradation by nucleases and poor membrane permeability which free phosphodiester oligonucleotides are subject to, but leaves unresolved the inefficiency of encapsulation of oligonucleotides in liposomes. We have coupled oligonucleotides to cholesterol via a reversible disulfide bond. This modification of oligonucleotides improved their association with immunoliposomes by a factor of about 10 in comparison to unmodified oligonucleotides. The presence of cholesteryl-modified oligonucleotides incorporated in the bilayer of liposomes did not interfere with the coupling of the targeting protein to the liposome surface. Free or cholesterol coupled oligonucleotides associated with liposomes and directed against the tat gene of HIV-1 were tested for inhibition of HIV-1 proliferation in acutely infected cells. We demonstrate that the cholesteryl-modified as well as unmodified oligonucleotides acquire the target specificity of the antibody on the liposome. Their antiviral activity when delivered into cells is sequence-specific. The activity of these modified or unmodified oligonucleotides to inhibit the replication of HIV was the same on an equimolar basis (EC50 around 0.1 microM). Cholesterol coupled oligonucleotides thus offer increased liposome association without loss of antiviral activity.
Sigma-Aldrich
Thiocholesterol