MilliporeSigma
  • Desaturation, dioxygenation, and monooxygenation reactions catalyzed by naphthalene dioxygenase from Pseudomonas sp. strain 9816-4.
Desaturation, dioxygenation, and monooxygenation reactions catalyzed by naphthalene dioxygenase from Pseudomonas sp. strain 9816-4.
Journal of bacteriology (1995-05-01)
D T Gibson, S M Resnick, K Lee, J M Brand, D S Torok, L P Wackett, M J Schocken, B E Haigler
ABSTRACT
The stereospecific oxidation of indan and indene was examined with mutant and recombinant strains expressing naphthalene dioxygenase of Pseudomonas sp. strain 9816-4. Pseudomonas sp. strain 9816/11 and Escherichia coli JM109(DE3)[pDTG141] oxidized indan to (+)-(1S)-indanol, (+)-cis-(1R,2S)-indandiol, (+)-(1S)-indenol, and 1-indanone. The same strains oxidized indene to (+)-cis-(1R,2S)-indandiol and (+)-(1S)-indenol. Purified naphthalene dioxygenase oxidized indan to the same four products formed by strains 9816/11 and JM109(DE3)[pDTG141]. In addition, indene was identified as an intermediate in indan oxidation. The major products formed from indene by purified naphthalene dioxygenase were (+)-(1S)-indenol and (+)-(1R,2S)-indandiol. The results show that naphthalene dioxygenase catalyzes the enantiospecific monooxygenation of indan to (+)-(1S)-indanol and the desaturation of indan to indene, which then serves as a substrate for the formation of (+)-(1R,2S)-indandiol and (+)-(1S)-indenol. The relationship of the desaturase, monooxygenase, and dioxygenase activities of naphthalene dioxygenase is discussed with reference to reactions catalyzed by toluene dioxygenase, plant desaturases, cytochrome P-450, methane monooxygenase, and other bacterial monooxygenases.
Sigma-Aldrich
2,3-Dihydrobenzofuran, 99%
Sigma-Aldrich
Indan, 95%
Supelco
Indan, analytical standard
Sigma-Aldrich
(R)-(−)-1-Indanol, 99%