MilliporeSigma
  • Alteration of spleen lymphocyte populations in rats with arthritis induced by muramyl dipeptide analogue or complete adjuvant.
Alteration of spleen lymphocyte populations in rats with arthritis induced by muramyl dipeptide analogue or complete adjuvant.
International journal of tissue reactions (1995-01-01)
T Sugawara, M Miyamoto, S Takada, M Nomura, M Kato
ABSTRACT
To examine the involvement of lymphocytes in the development of MDP-Lys(L18)-induced arthritis (MIA) in rats and the exacerbation of MIA by cyclosporin A (CsA), we analysed the spleen lymphocyte subset using monoclonal antibodies and flow cytometry during the development of arthritis and compared the results with those found in adjuvant-induced arthritis (AIA). Subcutaneous injection of MDP-Lys(L18) 4 mg/kg to male Lewis rats for 14 days caused very slight and quite clear increases in tarsal joint thickness on days 8 and 15, respectively. This increase was significantly enhanced by co-administration of CsA 10 mg/kg on both of these days. Adjuvant intracutaneously injected once increased the thickness only on day 15, and this was completely inhibited by CsA. The populations of CD4+ and CD8+ cells were increased and decreased, respectively, increasing the CD4+/CD8+ ratio, from day 8 in MIA. CsA enhanced the MDP-Lys(L18)-induced changes in these populations and caused additional decreases in the number of CD5+ cells. Only the CD4+ cell population was increased on day 15 in AIA, and this increase was inhibited by CsA. These results suggest that the spleen lymphocyte subsets in MIA have a different role from those in AIA, and that the contribution of enhancement of the subset changes to the exacerbating effect of CsA on MIA.